• USA Home
  • P2338 - Phosphoglucose Isomerase from baker's yeast (S. cerevisiae)

EMAIL THIS PAGE TO A FRIEND
P2338 Sigma

Phosphoglucose Isomerase from baker's yeast (S. cerevisiae)

lyophilized powder, 400-600 units/mg protein

Synonym: D-Glucose-6-phosphate ketol-isomerase, PGI, Phosphosaccharomutase

Purchase

Properties

Related Categories 5.3.x.x Intramolecular oxidoreductases, 5.x.x.x Isomerases, Biochemicals and Reagents, Cell Biology, Enzyme Class Index,
form   lyophilized powder
composition   Protein, ~50% modified Warburg-Christian
storage temp.   −20°C

Description

Unit Definition

One unit will convert 1.0 μmole of D-fructose 6-phosphate to D-glucose 6-phosphate per min at pH 7.4 at 25 °C.

Physical form

Essentially sulfate-free, powder containing carbohydrate and citrate buffer salts

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?