• USA Home
  • P2347 - Potassium sodium tartrate tetrahydrate

EMAIL THIS PAGE TO A FRIEND
P2347 Sigma

Potassium sodium tartrate tetrahydrate

meets USP testing specifications

Synonym: L(+)-Tartaric acid potassium Sodium salt, Rochelle salt, Seignette salt

  • CAS Number 6381-59-5

  • Linear Formula KOCOCH(OH)CH(OH)COONa · 4H2O

  • Molecular Weight 282.22

  •  Beilstein Registry Number 6113568

  •  EC Number 206-156-8

  •  MDL number MFCD00150989

  •  PubChem Substance ID 329820133

  • Popular Documents:  FTIR (PDF)  

Purchase

Properties

Related Categories Analytical/Chromatography, Biochemicals and Reagents, Chelators, Pharmacopoeia, Pharmacopoeia A-Z,
InChI Key   VZOPRCCTKLAGPN-ZFJVMAEJSA-L
assay   99%
suitability   meets USP testing specifications

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?