• USA Home
  • P2402 - Pro-His-Pro-Phe-His-Phe-Phe-Val-Tyr-Lys

EMAIL THIS PAGE TO A FRIEND
P2402 Sigma

Pro-His-Pro-Phe-His-Phe-Phe-Val-Tyr-Lys

≥97% (HPLC)

Purchase

Properties

assay   ≥97% (HPLC)
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?