• USA Home
  • P2418 - Pneumocystis TISSUE-TROL

EMAIL THIS PAGE TO A FRIEND
P2418 Sigma

Pneumocystis TISSUE-TROL

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, TISSUE-TROL Control Slides More...
biological source   from human lung tissue containing Pneumocystis carinii
shelf life   Expiry date on label
IVD   for in vitro diagnostic use

Description

Components

25 slides per box. 1 stained slide is provided with each box. The stained slide has been stained with Sigma Procedure No. HT100.

Application

Suitable for use with Sigma Silver Stain Procedure No. HT100

Other Notes

Due to raw material availability difficulties, the Pneumocystis Tissue-Trol is on indefinite back-order.

Legal Information

TISSUE-TROL is a trademark of Sigma-Aldrich Co. LLC

Price and Availability


All labs need water
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?