• USA Home
  • P2507 - 5α-Pregnane-3α,17α,20β-triol

EMAIL THIS PAGE TO A FRIEND
P2507 Sigma

5α-Pregnane-3α,17α,20β-triol

Synonym: 3α,17α,20β-Trihydroxy-5α-pregnane, Allopregnane-3α,17α,20β-triol

  • CAS Number 13933-75-0

  • Empirical Formula (Hill Notation) C21H36O3

  • Molecular Weight 336.51

  •  EC Number 237-708-6

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Biomarkers and Metabolomics Standards, Hormone Standards
InChI Key   SCPADBBISMMJAW-MUHHAPIYSA-N
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?