• USA Home
  • P2539 - Poly(guanylic-uridylic) acid potassium Sodium salt

EMAIL THIS PAGE TO A FRIEND
P2539 Sigma

Poly(guanylic-uridylic) acid potassium Sodium salt

Synonym: Poly(G,U)

Purchase

Properties

feed ratio   G:U (1:1)
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?