EMAIL THIS PAGE TO A FRIEND
P2625 Sigma

D-(−)-Pantolactone

Synonym: (R)-(−)-α-Hydroxy-β,β-dimethyl-γ-butyrolactone, (R)-(−)-β,β-Dimethyl-α-hydroxy-γ-butyrolactone, (R)-(−)-Pantolactone, Pantoic acid γ-lactone

  • CAS Number 599-04-2

  • Empirical Formula (Hill Notation) C6H10O3

  • Molecular Weight 130.14

  •  Beilstein Registry Number 80957

  •  EC Number 209-963-3

  •  MDL number MFCD00005392

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

InChI Key   SERHXTVXHNVDKA-BYPYZUCNSA-N
bp   120-122 °C/15 mmHg(lit.)
mp   91 °C(lit.)

Description

Other Notes

Degradation product of pantothenic acid.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

237817 D-(−)-Pantolactone, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?