EMAIL THIS PAGE TO A FRIEND
P265 Sigma

Penitrem A

solid

Synonym: (5H,7H)-triol, 12-Chloro-3,3a,6a,8,9,9a,10,11,14,14b,14c,15,16,16a-tetradecahydro-14b,14c,17,17-tetramethyl-10-methylene-2-(1-methylethenyl)-7,8-(epoxymethano)-2H,6H-cylcobuta[5,6]benz[1,2-e]oxireno[4′,4′a]-1-benzopyrano[5′,6′:6,7]indeno[1,2-b]indole-3,4b,7d, Tremortin A

Purchase

Properties

form   neat
  solid
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
color   white
solubility   methanol: soluble10 mg/mL
format   neat
storage temp.   −20°C

Description

Biochem/physiol Actions

Selective blocker of high conductance Ca2+-activated (Maxi-K) potassium channels.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2811 6.1/PG 2
WGK Germany 
3
RTECS 
RY7535000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

P3053 Penitrem A, ≥95% (HPLC and TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?