• USA Home
  • P266 - Phorbol 12-phenylacetate 13-acetate 20-homovanillate

EMAIL THIS PAGE TO A FRIEND
P266 Sigma

Phorbol 12-phenylacetate 13-acetate 20-homovanillate

solid

Synonym: PPAHV

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Neurobiology, Neuroscience, Pharmacologicals,
InChI Key   OGRGWTXWAZBJKF-JXWJAAHMSA-N
form   solid
color   white
solubility   DMSO: soluble
  H2O: insoluble

Description

Biochem/physiol Actions

Vanilloid receptor agonist.

Legal Information

Sold in France, Germany, Great Britain and Switzerland under license, restricted to research use only, from Procyon Pharmaceuticals, Inc.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?