EMAIL THIS PAGE TO A FRIEND
P2676 Sigma

PDX G.F.

50-150 μm (dry), fractionation range 1000-5000 Da (globular proteins)

Purchase

Properties

Related Categories Gel Filtration Chromatography, Gel Filtration Media, Molecular Biology, Protein Chromatography, Proteomics More...
swelling   1 g swells to 4-6 mL
particle size   50-150 μm (dry)
pore size   1000-5000 Da fractionation range (globular proteins)

Description

Application

For Gel Filtration Chromatography

Other Notes

Cross-linked dextran beads

Legal Information

PDX is a trademark of Polydex Biologicals

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?