• USA Home
  • P2804 - Pectin Lyase Acidic enzyme from Aspergillus japonicus

EMAIL THIS PAGE TO A FRIEND
P2804 Sigma

Pectin Lyase Acidic enzyme from Aspergillus japonicus

buffered aqueous glycerol solution, 50-150 units/mg protein, pH 5.0 (Lowry, assayed in 100 mM citrate-phosphate buffer)

Synonym: Poly(methoxygalacturonide) lyase

Purchase

Properties

form   buffered aqueous glycerol solution
foreign activity   cellulase, hemicellulase and glycosaminidase activities, essentially free
  polygalacturonase <10 units/mg protein
storage temp.   2-8°C

Description

Preparation Note

Purified from pectolyase

Unit Definition

One unit will cause a ΔA235 of 1.0 per min at 40°C due to the release of unsaturated products from Pectin (P 9135).

Physical form

Solution in 50% glycerol containing 40 mM sodium acetate buffer, pH 4.5, and 1 mM PMSF

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?