• USA Home
  • P2892 - 1,2-Dihexadecanoyl-rac-glycero-3-phospho-rac-(1-glycerol) ammonium salt

EMAIL THIS PAGE TO A FRIEND
P2892 Sigma

1,2-Dihexadecanoyl-rac-glycero-3-phospho-rac-(1-glycerol) ammonium salt

~99% (TLC)

Synonym: DL-α-Phosphatidyl-DL-glycerol, dipalmitoyl ammonium salt, L-α-Phosphatidyl-DL-glycerol, dipalmitoyl

Purchase

Properties

InChI Key   KSVUSCAQEBSOIJ-UHFFFAOYSA-N
assay   ~99% (TLC)
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

P9789 1,2-Dipalmitoyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt, ≥99% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?