• USA Home
  • P2936 - Thermocouple probe, PTFE coated stainless steel
EMAIL THIS PAGE TO A FRIEND
P2936 Sigma
Thermocouple probe, PTFE coated stainless steel
Type J
Purchase
Properties
Related Categories Barnant, Barnant Temperature Measurement, Labware, Manufacturer Browser, Temperature,
mfr. no.   Barnant
parameter   -250-260 °C temp. range (-418-500 °F)
L × diam.   10 in. × 0.145 in.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
related product
Product #
Description
Add to Cart
92451 Timestrip Plus 25 °C
08168 Timestrip Plus 8 °C
06693 Timestrip Plus -20 °C
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?