• USA Home
  • P2975 - Monoclonal Anti-PAG608 (p53-induced protein) antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
P2975 Sigma

Monoclonal Anti-PAG608 (p53-induced protein) antibody produced in mouse

clone 608-45, ascites fluid

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Cell Cycle, Antibodies to Nuclear Proteins,
species reactivity   rat, human, mouse
application(s)   immunocytochemistry: suitable
  indirect ELISA: suitable
  western blot: 1:500 using a whole cell extract of temperature-sensitive p53-induced cultured cell line
clone   608-45, monoclonal
antibody form   ascites fluid
isotype   IgG1
contains   15 mM sodium azide
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... ZMAT3(64393)
mouse ... Zmat3(22401)
biological source   mouse
conjugate   unconjugated

Description

Immunogen

GST fusion protein containing the C-terminal 59 residues of rat PAG608.

Specificity

Reacts specifically with PAG608. Recognizes an epitope present at the C-terminal 59 residues of the PAG608 molecule. Stains a strong band at approx. 25 kDa, representing a truncated protein, which probably stems from an alternative translation start from the Met21 of the PAG608 molecule and a weak band at 34 kDa. In some cell lines (e.g., NIH 3T3) a signal of approx. 20 kDa may also be detected.

Price and Availability


Antibody Explorer

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?