• USA Home
  • P3039 - Peroxidase Anti-Peroxidase Soluble Complex antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
P3039 Sigma

Peroxidase Anti-Peroxidase Soluble Complex antibody produced in mouse

affinity isolated antibody, buffered aqueous solution

Synonym: PAP

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, PAP, PD-PG, Primary Antibodies,
biological source   mouse
antibody form   affinity isolated antibody
form   buffered aqueous solution
application(s)   immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:200
shipped in   dry ice
storage temp.   −20°C

Description

Physical form

Solution in 8 mM sodium acetate, 15 mM ammonium acetate buffered saline, pH 7.2, containing 0.5% bovine serum albumin and 0.01% thimerosal.

Application

Sigma PAP is assayed by immunoelectrophoresis, Ouchterlony double diffusion, and immunohistochemical staining techniques to ensure purity and functional reactivity. It is recommended that the optimal working dilution be determined in the specific assay system.

Biochem/physiol Actions

The Peroxidase Anti-Peroxidase (PAP) soluble complex is prepared by a modification of the method of Sternberger using specific polyclonal or monoclonal antibodies to horseradish peroxidase and highly purified enzyme. Since the enzyme is not covalently coupled to the antibody, the soluble complex retains its enzyme and antibody reactivity with distinct advantages over enzyme-conjugated antibodies.

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

How to Choose a Secondary Antibody

The following information is provided to help you decide which secondary antibody may be best for your particular application.
Keywords: Amplification, Enzyme-linked immunosorbent assay, Flow cytometry, Immobilization, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western blot

Secondary Antibodies, Conjugates and Kits

Secondary antibodies are polyclonal or monoclonal antibodies that bind to primary antibodies or antibody fragments, such as the Fc or Fab regions. They are typically labeled with probes that make the...
Keywords: Absorption, Adsorption, Amplification, Bacterial conjugations, Cancer, Digestions, Electrophoresis, Enzyme-linked immunosorbent assay, Flow cytometry, Gel electrophoresis, Gene expression, Hormones, Immobilization, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunology, Immunoprecipitation, Infrared spectroscopy, Microbiology, Microscopy, Purification, Vitamins, Western blot

Related Content

Antibody Explorer - Over 80,000 Products

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?