• USA Home
  • P3063 - Immobilon®-PSQ Polyvinylidene difluoride membranes

EMAIL THIS PAGE TO A FRIEND
P3063 Sigma

Immobilon®-PSQ Polyvinylidene difluoride membranes

size 10 cm × 10 cm

Synonym: PVDF

Purchase

Description

General description

Immobilon-PSQ membranes are PVDF with greater protein binding capacity than standard Immobilon-P. Thus, in protein sequencing applications, dramatically higher yields can be obtained. Repetitive yields are typically higher than 90%.

Other Notes

Note: We recommend traditional Immobilon-P membranes for routine Western blotting applications. Immobilon-PSQ membranes are functional in these applications but will have higher background.

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?