• USA Home
  • P3117 - Monoclonal Anti-p73α antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
P3117 Sigma

Monoclonal Anti-p73α antibody produced in mouse

~1 mg/mL, clone ER-13, purified immunoglobulin, buffered aqueous solution

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Cell Cycle, Antibodies to Nuclear Proteins,
species reactivity   human, monkey
application(s)   direct immunofluorescence: suitable
  immunoprecipitation: 2 μg using 1 mg protein lysate
  western blot: 2 μg/mL using COS1 monkey kidney cells
  western blot: 2 μg/mL using IM9 human myeloma cell lysates
clone   ER-13, monoclonal
concentration   ~1 mg/mL
antibody form   purified immunoglobulin
form   buffered aqueous solution
isotype   IgG1
mol wt   antigen mol wt ~80 kDa
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... TP73(7161)
biological source   mouse
conjugate   unconjugated

Description

Immunogen

amino acids 380-637 from purified monkey recombinant p73α.

Physical form

Solution in phosphate buffered saline pH 7.4, containing 0.08% sodium azide.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Documents

Certificate of Analysis

Certificate of Origin

Safety Information

WGK Germany  -
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?