• USA Home
  • P3147 - Phosphatase, Acid, Prostatic from bovine semen

EMAIL THIS PAGE TO A FRIEND
P3147 Sigma

Phosphatase, Acid, Prostatic from bovine semen

lyophilized powder, ~2 units/g solid

Purchase

Description

Other Notes

Note: 60 Sigma units = 1 μmolar unit.

Unit Definition

One unit will hydrolyze 1.0 μmole of p-nitrophenyl phosphate per min at pH 4.8 at 37 °C.

Application

Acid phosphatase is routinely used to dephosphorylate proteins and nucleic acids. Acid phosphatase is an acid hydrolase that normally resides in lysosomes. It is used as marker for the identification of lysosomes in subcellular fractionations. Prostatic acid phosphatase has been used to study the stereochemical course of phospho group .

Biochem/physiol Actions

Acid phosphatase is a cytoplasmic lysosomal hydrolase that catalyzes the conversion of an orthophosphoric monoester and water to an alcohol and orthophosphate. Prostatic acid phosphatase activity is the difference between the total acid phosphatase activity and the acid phosphatase activity in the presence of 20 mM tartrate.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?