• USA Home
  • P3223 - (−29) M13 Reverse Primer Set

EMAIL THIS PAGE TO A FRIEND
P3223 Sigma

(−29) M13 Reverse Primer Set

Synonym: 5′-CAG GAA ACA GCT ATG ACC-3′

Purchase

Description

Components

Set of four vials of primer, each individually labeled at the 5′-end with:
FAM: (5[6]-carboxyfluorescein)
JOE: (6-carboxy-4′,5′-dichloro-2′,7′-dimethoxyfluorescein)
ROX: (5[6]-carboxy-X-rhodamine), and
TAMRA: (5[6]-carboxytetramethylrhodamine.
Supplied as solutions, 50 μl each, in tris-EDTA buffer, pH 7.5, at 1 picomole/μl

Analysis Note

Tested for purity with HPLC, PAGE, and OD.

Packaging

Packaged in amber tubes to protect from light.

Application

(−29) M13 Reverse Primer Set is a single-stranded oligonucleotide with 5′-hydroxyl and 3′-hydroxyl ends and a selection of four fluorescent lables for use in polymerase chain reaction protocols (PCR).

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?