• USA Home
  • P3226 - Protein L-Peroxidase from Peptostreptococcus magnus

EMAIL THIS PAGE TO A FRIEND
P3226 Sigma

Protein L-Peroxidase from Peptostreptococcus magnus

recombinant, expressed in E. coli, buffered aqueous glycerol solution

Purchase

Properties

Related Categories Antibodies, Protein A, G and L Detection Reagents, Secondary Antibodies More...
recombinant   expressed in E. coli
form   buffered aqueous glycerol solution
human IgG binding affinity   2-3 × 109 M-1
application(s)   direct ELISA: 1:3,000
  dot blot: suitable
  western blot: suitable
solubility   PBS, pH 7.2: soluble (May be diluted in buffer)
conjugate   peroxidase conjugate
shipped in   wet ice
storage temp.   −20°C

Description

Biochem/physiol Actions

Protein L binds immunoglobulins from a variety of sources primarily through κ light chain interactions without interfering with the antigen binding site of Igs. This means that Protein L binds to a wider range of Ig classes and subclasses from a variety of species than any other commercially available Ig binding protein. It contains four Ig binding domains (B-domains). Protein L does not bind to bovine, sheep or goat immunoglobulins. Protein L-Peroxidase Conjugate may be used for the detection of IgG, IgM, IgA, IgE and IgD containing κ light chains. It may be used to detect Fab and scFv fragments containing κ light chains.

Preparation Note

Prepared with recombinant protein L conjugated to horseradish peroxidase using glutaraldehyde coupling chemistry.

Specificity

Binds Igs with κ light chain only. Does not bind λ light chain.

Physical form

Solution in 0.01 mM phosphate bufferered saline, pH 7.2, containing 50% glycerol.

Price and Availability


Antibody Explorer

Explore with Confidence

Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

How to Choose a Secondary Antibody

The following information is provided to help you decide which secondary antibody may be best for your particular application.
Keywords: Amplification, Enzyme-linked immunosorbent assay, Flow cytometry, Immobilization, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western blot

Secondary Antibodies, Conjugates and Kits

Secondary antibodies are polyclonal or monoclonal antibodies that bind to primary antibodies or antibody fragments, such as the Fc or Fab regions. They are typically labeled with probes that make the...
Keywords: Absorption, Adsorption, Amplification, Bacterial conjugations, Cancer, Digestions, Electrophoresis, Enzyme-linked immunosorbent assay, Flow cytometry, Gel electrophoresis, Gene expression, Hormones, Immobilization, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunology, Immunoprecipitation, Infrared spectroscopy, Microbiology, Microscopy, Purification, Vitamins, Western blot

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?