EMAIL THIS PAGE TO A FRIEND
P3240 Sigma

Peganole

95%, solid

  • Empirical Formula (Hill Notation) C11H12N2O

  • Molecular Weight 188.23

  •  MDL number MFCD00236491

  •  PubChem Substance ID 329820165

Purchase

Description

Biochem/physiol Actions

Acetylcholinestearase inhibitor; psychotomimetic agent in dogs and cats.

Caution

Protect from light.

Price and Availability


Neuroscience Grant Programm
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?