• USA Home
  • P3272 - Phenoxyacetylcellulose

EMAIL THIS PAGE TO A FRIEND
P3272 Sigma

Phenoxyacetylcellulose

Purchase

Description

General description

Phenoxyacetylcellulose is a support for the immobilization of enzymes and other proteins by hydrophobic adsorption.

Application

Phenoxyacetylcellulose is used to immobilize enzymes. Phenoxyacetylcellulose traps have been used to remove the enzyme glucose oxidase by physical adsorption in a study assessing catalytic determinations of enzymes. It has also been used in a study to investigate bioelectrochemical detection systems for substrates of dehydrogenases.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?