• USA Home
  • P3293 - Phe-Leu-Phe-Gln-Pro-Gln-Arg-Phe amide

EMAIL THIS PAGE TO A FRIEND
P3293 Sigma

Phe-Leu-Phe-Gln-Pro-Gln-Arg-Phe amide

≥95% (HPLC)

Synonym: F-8-F-NH2

  • CAS Number 99566-27-5

  • Empirical Formula (Hill Notation) C54H76N14O10

  • Molecular Weight 1081.27

  •  MDL number MFCD00076704

  •  PubChem Substance ID 24897882

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Other Peptides, Peptides and Proteins, Peptides for Cell Biology More...
InChI Key   HWYCFZUSOBOBIN-AQJXLSMYSA-N
assay   ≥95% (HPLC)
storage temp.   −20°C
Gene Information   rat ... Npffr1(64107)

Description

Biochem/physiol Actions

Attenuates analgesic effects of morphine

Other Notes

Endogenous peptide found in periaqueductal grey and in dorsal spinal cord

Price and Availability


Amplified Detection

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?