• USA Home
  • P3315 - Polyoxyethylene (8) stearate

EMAIL THIS PAGE TO A FRIEND
P3315 Sigma

Polyoxyethylene (8) stearate

Synonym: Myrj 45

Purchase

Properties

Related Categories Biochemicals and Reagents, Detergents, Detergents N to Z, Non-Ionic Detergents
description   non-ionic
InChI Key   WPXCYCTVMORDCF-UHFFFAOYSA-N

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
MD0907300

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?