EMAIL THIS PAGE TO A FRIEND
P3329 Sigma

Poly-L-isoleucine

mol wt 5,000-15,000

Purchase

Properties

Related Categories Amino Acids, Biochemicals and Reagents, Homopolymers, Polyamino Acids
mol wt   mol wt 5,000-15,000
storage temp.   −20°C

Description

Analysis Note

Molecular weight based on viscosity.

Other Notes

For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?