• USA Home
  • P3363 - Protein Sequencing and Identification Using Tandem Mass Spectrometry
EMAIL THIS PAGE TO A FRIEND
P3363 Sigma
Protein Sequencing and Identification Using Tandem Mass Spectrometry
  •  ISBN-10 0-471-32249-0
  •  ISBN-13 978-0-471-32249-8
Purchase
Properties
Related Categories Books and Software, Labware, Proteomics Books, Spectroscopy Books
publication info   M. Kinter and N.E. Sherman, Wiley-Interscience, 2001, 301 pp., hard cover
mfr. no.   Wiley
Description
General description
Readers will find in this book complete experimental detail to apply mass spectrometry techniques in their laboratoriess and learn about the proper selection and operation of instruments. Coverage includes peptide fragmentation and interpretation of product ion spectra, preparation of protein digests for sequencing experiments, mass spectrometric analysis using capillary liquid chromatography, characterization of modified peptides using tandem mass spectrometry and techniques for protein identification by database searches.
Table of Contents
An Introduction to Protein Sequencing Using Tandem Mass Spectrometry.
The Primary Structure of Proteins and a Historical Overview of Protein Sequencing.
Fundamental Mass Spectrometry.
Collisionally Induced Dissociation of Protonated Peptide Ions and the Interpretation of Product Ion Spectra.
Basic Polyacrylamide Gel Electrophoresis.
The Preparation of Protein Digests for Mass Spectrometric Sequencing Experiments.
Mass Spectrometric Analysis of Tryptic Digests.
Protein Identification by Database Searching.
Sequence Analysis of Novel Proteins.
The Characterization of Post-Translationally Modified Proteins Using Tandem Mass Spectrometry.
Index.
Price and Availability
Detect Your Unseen Protein
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?