• USA Home
  • P3364 - PurePak PCR microtubes
EMAIL THIS PAGE TO A FRIEND
P3364 Sigma
PurePak PCR microtubes
volume 0.5 mL, 0.5 mL PCR tubes, nuclease and pyrogen-free
Purchase
Description
General description
Reaching into a bulk bag of tubes can cause contamination; PurePak packaging solves this problem by dividing tubes into ten separate PurePaks. PurePaks can be opened as needed to protect unused tubes from contamination. Thin walled tubes are precision-molded with premium, non-wettable polypropylene and receive multi-point, quality inspections to ensure unsurpassed performance. Certified Rnase-, DNase- and pyrogen-free.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Protocols & Articles
Protocols
Amplification of DNA Using Jumpstart REDTaq DNA Polymerase D8187 Protocol
Note: The use of DMSO or formamide with JumpStart REDTaq DNA Polymerase is not recommended due to interference with the enzyme-antibody complex. Other co-solvents, solutes (e.g., salts) and extremes ...
Keywords: Amplification, Evaporation, Nucleic acid annealing, Nucleic acid denaturation, Polymerase chain reaction, Reductions, Size-exclusion chromatography, Solvents, Titrations
Amplification of DNA Using Jumpstart Taq DNA Polymerase D9307 Protocol
Note: JumpStart Taq DNA polymerase has been shown to work effectively with up to 5% v/v DMSO. Other co-solvents, solutes (e.g., salts) and extremes in pH or other reaction conditions may reduce the a...
Long and Accurate PCR Amplification of DNA D8045 Protocol
Effective denaturation is accomplished by the use of higher temperatures for shorter periods of time or by the use of co-solvents, such as dimethyl sulfoxide. Addition of DMSO in the reaction at a fi...
Long and Accurate PCR Amplification of DNA with RedAccuTaq D4812 Protocol
Reaction Optimization Reliable amplification of long DNA sequences requires: 1) effective denaturation of DNA template, 2) adequate extension times to produce large products and 3) protection of targ...
Low Contaminant Amplification of DNA Using MTP Taq DNA Polymerase D7442 Protocol
Every precaution should be taken to avoid contamination of reagents with unknown/unwanted DNA. This includes the following:
Reduce Non-specific PCR Amplification- Jumpstart Taq Polymerase D4148 Protocol
Note: JumpStart Taq DNA polymerase has been shown to work effectively with up to 5% v/v DMSO. Other co-solvents, solutes (e.g., salts) and extremes in pH or other reaction conditions may reduce the a...
Keywords: AGE, Amplification, Electrophoresis, Evaporation, Gel electrophoresis, Nucleic acid annealing, Nucleic acid denaturation, Polymerase chain reaction, Reductions, Size-exclusion chromatography, Solvents, Titrations
Reverse Transcription Using ReadyScript cDNA Synthesis RDRT Protocol
Preparation Place components on ice. Mix, and then briefly centrifuge to collect contents at the bottom of the tube
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?