• USA Home
  • P3400 - 1,2-Dipalmitoyl-rac-glycero-3-phospho(dimethylaminoethanol)

EMAIL THIS PAGE TO A FRIEND
P3400 Sigma

1,2-Dipalmitoyl-rac-glycero-3-phospho(dimethylaminoethanol)

~99%

Synonym: DL-α-Phosphatidylethanolamine, dipalmitoyl, N,N-dimethyl

  • CAS Number 54285-59-5

  • Empirical Formula (Hill Notation) C39H78NO8P

  • Molecular Weight 720.01

  •  MDL number MFCD00042876

  •  PubChem Substance ID 24898406

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Biology, Glycerophosphoethanolamines, Glycerophospholipid Metabolism, Glycerophospholipids,
InChI Key   SKWDCOTXHWCSGS-UHFFFAOYSA-N
assay   ~99%
storage temp.   −20°C

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?