• USA Home
  • P3428 - Endoproteinase Lys-C from Lysobacter enzymogenes

EMAIL THIS PAGE TO A FRIEND
P3428 Sigma

Endoproteinase Lys-C from Lysobacter enzymogenes

suitable for protein sequencing, lyophilized powder

Purchase

Description

Unit Definition

One unit will hydrolyze 1.0 μmole of N-p-tosyl-Gly-Pro-Lys p-nitroanilide per min at pH 7.7 at 25 °C.

Price and Availability


SILu Mab K1 IgG 1k Mass Spec Standard

Detect Your Unseen Protein

New Internal Standards for Mass Spectrometry
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Stable Isotope-labeled MS Internal Standards

What happens when you combine state-of-the-art technology with scientific prowess? Full-length protein standards with the highest purity and isotopic incorporation in the industry. Stable isotope-lab...
Keywords: Atomic absorption spectroscopy, Digestions, Mass spectrometry, Phosphorylations

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?