• USA Home
  • P3453 - O-Phospho-L-threonine Agarose

EMAIL THIS PAGE TO A FRIEND
P3453 Sigma

O-Phospho-L-threonine Agarose

lyophilized powder

Purchase

Properties

form   lyophilized powder
extent of labeling   5-15 μmol per mL
matrix   cross-linked 4% beaded agarose
matrix activation   cyanogen bromide
matrix attachment   amino
matrix spacer   1 atom
storage temp.   −20°C

Description

Preparation Note

Stabilizer with lactose

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?