• USA Home
  • P3544 - 1-Palmitoyl-3-stearoyl-rac-glycerol

EMAIL THIS PAGE TO A FRIEND
P3544 Sigma

1-Palmitoyl-3-stearoyl-rac-glycerol

~99%

Synonym: 1-Hexadecanoyl-3-octadecanoyl-rac-glycerol

Purchase

Properties

Related Categories Biochemicals and Reagents, Diacylglycerols, Glycerolipids, Lipids
InChI Key   BFTGWUUHOMAGPO-UHFFFAOYSA-N
assay   ~99%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?