EMAIL THIS PAGE TO A FRIEND
P3567 Sigma

Anti-CD45R, Mouse

clone RA3-6B2, purified immunoglobulin

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, CD0-CDC, Primary Antibodies
antibody form   purified immunoglobulin
clone   RA3-6B2, monoclonal
species reactivity   mouse
isotype   IgG2a
conjugate   unconjugated
shipped in   wet ice
storage temp.   2-8°C

Description

Other Notes

CD45R is expressed on all B cells,
B cell precursors, and a subset of cytotoxic T lymphocytes.

Immunogen

Mouse pre-B tumor cells.

Preparation Note

Prepared by conjugation to R-phycoerythrin (PE). This orange-red dye is efficiently excited at 488 nm and emits at 578 nm.

Specificity

Recognizes the Ly5, B220 or 220 kDaisoform of the CD45 cell surface molecule. This clone recognizes only the B cell determinant of all strains of mice tested.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?