• USA Home
  • P3631 - Phenyl N-acetyl-β-D-glucosaminide

EMAIL THIS PAGE TO A FRIEND
P3631 Sigma

Phenyl N-acetyl-β-D-glucosaminide

≥98%

Synonym: Phenyl 2-acetamido-2-deoxy-β-D-glucopyranoside

Purchase

Properties

InChI Key   ZUJDLWWYFIZERS-DHGKCCLASA-N
assay   ≥98%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?