EMAIL THIS PAGE TO A FRIEND
P3693 Sigma

Phenyl-Agarose

saline suspension

Purchase

Properties

Related Categories Affinity Chromatography, Hydrophobic Interaction, Molecular Biology, Protein Chromatography, Proteomics More...
form   saline suspension
matrix   cross-linked 4% beaded agarose
matrix activation   epichlorohydrin
matrix attachment   hydroxyl
matrix spacer   3 atoms
capacity   10-25 mg/mL binding capacity (β-lactoglobulin)
  ≥20 mg/mL binding capacity (human serum albumin)
storage temp.   2-8°C

Description

Physical form

Suspension in 1.0 M NaCl containing 20% ethanol

Application

Phenyl-agarose is used in protein chromatography, affinity chromatography and hydrophobic interactions. Phenyl-agarose has been used to further the understanding of the digestive process of the pest species Dysdercus peruvianus as well as to study early onset dementia and Alzheimer’s disease.

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?