• USA Home
  • P3708 - Phe-Gln-Val-Val-Cys(NPYS)-Gly-NH2

EMAIL THIS PAGE TO A FRIEND
P3708 Sigma

Phe-Gln-Val-Val-Cys(NPYS)-Gly-NH2

≥95% (HPLC)

  • CAS Number 128102-74-9

  • Empirical Formula (Hill Notation) C34H48N10O9S2

  • Molecular Weight 804.94

  •  MDL number MFCD00238111

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Other Peptides, Peptides and Proteins, Peptides for Cell Biology More...
InChI Key   RUUXMQHJVXHBGZ-ZCBIBPQVSA-N
assay   ≥95% (HPLC)
storage temp.   −20°C

Description

Biochem/physiol Actions

Selectively blocks thrombin-induced platelet aggregation by binding to platelet calpain. Does not inhibit platelet aggregation induced by hemostasis factors other than thrombin.

Packaging

1, 5 mg in serum bottle

Price and Availability


Detect Your Unseen Protein

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?