• USA Home
  • P3716 - 1,2-Didecanoyl-sn-glycerol 3-phosphate sodium salt

EMAIL THIS PAGE TO A FRIEND
P3716 Sigma

1,2-Didecanoyl-sn-glycerol 3-phosphate sodium salt

≥99% (TLC)

Synonym: L-α-Phosphatidic acid, didecanoyl sodium salt

Purchase

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?