EMAIL THIS PAGE TO A FRIEND
P3741 Sigma

[Tyr783]-PLCγ

≥80% (HPLC), solid

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Other Peptides, Peptides and Proteins, Peptides for Cell Biology More...
assay   ≥80% (HPLC)
form   solid
mol wt   mol wt 1823
composition   Peptide content, ≥75%
solubility   H2O: soluble
storage temp.   −20°C

Description

Biochem/physiol Actions

Negative control peptide for the [pTyr783]-PLCγ rabbit polyclonal antibody.

Price and Availability


Amplified Detection

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?