• USA Home
  • P3777 - 1,2-Dihexadecyl-rac-glycero-3-phosphocholine

EMAIL THIS PAGE TO A FRIEND
P3777 Sigma

1,2-Dihexadecyl-rac-glycero-3-phosphocholine

≥99% (TLC)

Synonym: 1,2-Di-O-hexadecyl-rac-glycero-3-phosphocholine, rac-Phosphatidylcholine, 1,2-dihexadecyl, DL-β,γ-Dihexadecyl-α-lecithin

  • Empirical Formula (Hill Notation) C40H84NO6P

  • Molecular Weight 706.07

  •  Beilstein Registry Number 4168071

  •  EC Number 274-018-4

  •  MDL number MFCD00043213

  •  PubChem Substance ID 24898452

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Biology, Glycerophospholipid Metabolism, Glycerophospholipids, Glycerophosphonocholines,
InChI Key   DACAUPQETXAEBT-UHFFFAOYSA-N
assay   ≥99% (TLC)
storage temp.   −20°C

Price and Availability


The Future of Bioimaging

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?