• USA Home
  • P3798 - Pyruvate Dehydrogenase from Lactobacillus delbrueckii

EMAIL THIS PAGE TO A FRIEND
P3798 Sigma

Pyruvate Dehydrogenase from Lactobacillus delbrueckii

lyophilized powder, ≥1 units/mg solid

Purchase

Properties

Related Categories Cell Biology, General Metabolic Enzymes, General Metabolic Enzymes I-Z, Metabolomics
form   lyophilized powder
storage temp.   2-8°C

Description

Unit Definition

One unit will produce 1.0 μmole of acetyl phosphate and CO2 from pyruvate and inorganic phosphate per min at pH 6.3 at 37 °C.

Physical form

Lyophilized powder containing approx. 50% mannitol and FAD as stabilizers.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?