• USA Home
  • P3841 - L-α-Phosphatidylcholine

EMAIL THIS PAGE TO A FRIEND
P3841 Sigma

L-α-Phosphatidylcholine

~99%, from bovine brain, lyophilized powder

Synonym: 1,2-Diacyl-sn-glycero-3-phosphocholine, 3-sn-Phosphatidylcholine, L-α-Lecithin, Azolectin, PC

Purchase

Description

Biochem/physiol Actions

A major structural phospholipid in brain, comprising approx. 15% of total lipid; primarily localized to gray matter.

Packaging

packaged under argon

Preparation Note

Prepared chromatographically

Price and Availability


The Future of Bioimaging

All labs need water
Safety & Documentation

Safety Information

WGK Germany 
1
RTECS 
OG7565000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

P3556 L-α-Phosphatidylcholine, from egg yolk, Type XVI-E, ≥99% (TLC), lyophilized powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?