• USA Home
  • P3863 - 26S Proteasome Fraction from rabbit

EMAIL THIS PAGE TO A FRIEND
P3863 Sigma

26S Proteasome Fraction from rabbit

65%, buffered aqueous glycerol solution

Purchase

Description

Biochem/physiol Actions

Used for degradation of preformed ubiquitin-protein conjugates.

General description

Manufactured for Sigma by Boston Biochem., Inc.

Preparation Note

Partially purified 26S Proteasome fraction, free of conjugation enzymes E1, E2s, E3s.

Physical form

Solution contains 50 mM HEPES, PH 7.6, 1.0 mM MgATP, 1.0 mM DTT, 150 mM NaCl and 10% glycerol

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?