EMAIL THIS PAGE TO A FRIEND
P3896 Sigma

Piperazine Anhydrous

Purchase

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2579 8 / PGIII
WGK Germany 
1
RTECS 
TK7800000
Flash Point(F) 
228.2 °F
Flash Point(C) 
109 °C

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

P45907 Piperazine, ReagentPlus®, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?