• USA Home
  • P3909 - Pituitary acetone powder from salmon

EMAIL THIS PAGE TO A FRIEND
P3909 Sigma

Pituitary acetone powder from salmon

Purchase

Properties

Related Categories Acetone Powders, Biochemicals and Reagents, Proteins and Derivatives More...
storage temp.   −20°C

Description

Packaging

100 mg in poly bottle

Application

Pituitary acetone powder from salmon can be used to induce maturation and ovulation in fish.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?