• USA Home
  • P3950 - Lab-3 adjustable volume digital pipettes
EMAIL THIS PAGE TO A FRIEND
P3950 Sigma
Lab-3 adjustable volume digital pipettes
capacity 0.5-10 μL
Purchase
Properties
Related Categories Labnet, Labnet Pipettors, Labnet Pipettors & Tips, Labware, Liquid Handling Equipment,
mfr. no.   Labnet
availability   not available in EU
capacity   0.5-10 μL
Description
General description
Air-displacement pipettes with tip ejectors (except P 4575) and digital display of volume setting. Continuously adjustable within volume range.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?