• USA Home
  • P4002 - Phosphatase, Alkaline from porcine intestinal mucosa

EMAIL THIS PAGE TO A FRIEND
P4002 Sigma

Phosphatase, Alkaline from porcine intestinal mucosa

≥1 unit/mg solid (in glycine buffer)

Synonym: Orthophosphoric-monoester phosphohydrolase (alkaline optimum)

Purchase

Description

Unit Definition

One unit will hydrolyze 1.0 μmole of 4-nitrophenyl phosphate per minute at pH 10.4 at 37 ° C.

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

inhibitor

Product #

Description

Add to Cart

D6518 Diethylenetriaminepentaacetic acid, ≥99% (titration)
P9375 1,10-Phenanthroline monohydrate, reagent grade
E4378 Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
ED2SS Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)

substrate

Product #

Description

Add to Cart

N2770 SIGMAFAST p-Nitrophenyl phosphate Tablets, tablet, To prepare 20 mL
N1891 SIGMAFAST p-Nitrophenyl phosphate Tablets, tablet, To prepare 5 mL
M3168 4-Methylumbelliferyl phosphate (4-MUP) Liquid Substrate System, liquid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?