• USA Home
  • P4010 - Pyruvate Kinase–Polyacrylamide from rabbit muscle

EMAIL THIS PAGE TO A FRIEND
P4010 Sigma

Pyruvate Kinase–Polyacrylamide from rabbit muscle

20-30 units/g solid, pH 7.6, 30 °C

Purchase

Properties

contains   30% borate buffer salts
storage temp.   2-8°C

Description

Analysis Note

Protein determined by biuret.

Unit Definition

One unit will convert 1.0 μmole of phospho(enol)pyruvate to pyruvate per min at pH 7.6 at 37°C.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?