• USA Home
  • P4022 - pGlu-His-Gly amide trifluoroacetate salt hydrate

EMAIL THIS PAGE TO A FRIEND
P4022 Sigma

pGlu-His-Gly amide trifluoroacetate salt hydrate

≥97% (HPLC)

  • Empirical Formula (Hill Notation) C13H18N6O4 · xC2HF3O2 · yH2O

  • Molecular Weight 322.32 (anhydrous free base basis)

  •  PubChem Substance ID 24898484

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides and Tripeptides, Peptides, Tripeptides
InChI Key   USTKPIRPLXGRSE-CDEWPDHBSA-N
assay   ≥97% (HPLC)
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?