• USA Home
  • P4038 - Polycytidylic acid-Polyguanylic acid sodium salt

EMAIL THIS PAGE TO A FRIEND
P4038 Sigma

Polycytidylic acid-Polyguanylic acid sodium salt

Synonym: Poly(C) • Poly(G)

Purchase

Description

Other Notes

Double-stranded homopolymer

Application

Polycytidylic acid-Polyguanylic acid (polyGC), a synthetic polynucleotide, is a double stranded homodimer used for physicochemical analysis of RNA structure.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?