• USA Home
  • P4046 - Phosphodiesterase 3′:5′-cyclic nucleotide-activator−FITC labeled

EMAIL THIS PAGE TO A FRIEND
P4046 Sigma

Phosphodiesterase 3′:5′-cyclic nucleotide-activator−FITC labeled

>40,000 units/mg protein (Lowry)

Synonym: Calmodulin-FITC

Purchase

Properties

storage temp.   −20°C

Description

Unit Definition

One unit will stimulate 0.016 activated unit of phosphodiesterase, 3′:5′-cyclic nucleotide, P 0520, in a 3 ml reaction volume at pH 7.5 and 30°C, to 50% of the maximum activity of the enzyme when saturated with activator, in the presence of 0.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?