• USA Home
  • P4150 - N-(10-[1-Pyrene]dec-9-enoyl) cerebroside

EMAIL THIS PAGE TO A FRIEND
P4150 Sigma

N-(10-[1-Pyrene]dec-9-enoyl) cerebroside

90-95%

Purchase

Properties

assay   90-95%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?